Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Phenylenediamine

Catalog No. P239545G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
15 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P239545G 5 g
P23954500G 500 g
P2395415KG 15 kg
P239541KG 1 kg
P23954100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. P239545G Supplier Sigma Aldrich Supplier No. P239545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

m-Phenylenediamine

Specifications

CAS 108-45-2
Melting Point 64°C to 66°C
Boiling Point 282°C to 284°C
Molecular Formula C6H8N2
Linear Formula C6H4-1,3-(NH2)2
MDL Number MFCD00007799
Quantity 5 g
Synonym 1,3-Benzenediamine; 1,3-Diaminobenzene; 1,3-Phenylenediamine; MPDA
Molecular Weight (g/mol) 108.14
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.