Learn More
Drg International Inc M. Pneumoniae Igg Elisa
Supplier:  Drg International Inc EIA3499
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Mycoplasma pneumoniae in serum or plasma. Mycoplasms are extracellular parasites, especially on mucous membranes, which can cause infections in human, animals, plants, and cell cultures. Mycoplasma pneumoniae is primarily a respiratory pathogen (obligat) in human involving the nasopharynx, throat, trachea, bronchi, bronchioles, and alveoli. The DRG Mycoplasma pneumoniae IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Mycoplasma pneumoniae antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Mycoplasma pneumoniae-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
Catalog No. 50-194-7579