Learn More
Drg International Inc DRG Mycoplasma Pneumoniae IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3500R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Mycoplasma pneumoniae in serum or plasma. This product is for Research Use Only. The DRG Mycoplasma pneumoniae IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with Mycoplasma pneumoniae antigen. Pretreated specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7580