Learn More
Creative Biomart Recombinant Ad26.COV2.S vaccine antigen
Catalog No. 502084718
Encompass_Preferred
Ad26.COV2.S is a lead recombinant vaccine candidate that contains an adenovirus serotype 26 (Ad26) vector expressing a stabilized SARS-CoV-2 spike protein. The vaccine was created in collaboration wit
Catalog No. 50-208-4718 Supplier Creative Biomart Supplier No. MPP340
Ad26.COV2.S is a lead recombinant vaccine candidate that contains an adenovirus serotype 26 (Ad26) vector expressing a stabilized SARS-CoV-2 spike protein. The vaccine was created in collaboration wit