Learn More
M-Pro NPak™ Nasopharyngeal Aspiration Kit
Catalog No. 22065100
Change view
Click to view available options
Quantity:
10 Kits/Box
Catalog No. Quantity
22-065-100 10 Kits/Box
1 options
This item is not returnable. View return policy
Catalog No. 22-065-100 Supplier M-Pro Supplier No. SAK 01
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
The gold standard for collecting RSV, influenza, and pertussis specimens
  • Age calibrated, anatomically designed catheter aids healthcare professional with depth of insertion
  • Two options include control syringe with Luer-lock catheter or non-intrusive bulb aspirator
  • Syringe/bulb conveniently pre-filled with sterile saline
Specifications
Product Type Syringe Aspiration Kit
For Use With (Application) Influenza and RSV specimen collection
Includes Control syringe with Luer-lock catheter or non-intrusive bulb aspirator
Quantity 10 Kits/Box