Learn More
Lifeline Cell Technology M PROST HU LIFELINE KT
SDP
Catalog No. 50311907
Encompass_Preferred
Includes ProstaLife Basal Medium 485 mL, ProstaLife LifeFactors Kit. Optimized for the culture of human prostate epithelial cells. ProstaLife supports the growth of these cells over a period of at least 15 population doublings.
Catalog No. 50-311-907 Supplier Lifeline Cell Technology Supplier No. LL0041
Includes ProstaLife Basal Medium 485 mL, ProstaLife LifeFactors Kit. Optimized for the culture of human prostate epithelial cells. ProstaLife supports the growth of these cells over a period of at least 15 population doublings.