Learn More
Julabo USA Inc Each, M16x1m in-line Pt100 sensor
SDP
Catalog No. 507534127
Encompass_Preferred
Bath Accessories, Julabo, M+R in-line Pt100 sensor with M16x1 male fittings, 1.5 m cable (measurement and control in external system). M16x1 male fittings with hose barb connections (barb connections are removable). For in-line Pt100 monitoring/contol from an external position in the flow circuit. For use with JULABO systems having external Pt100 capability.
Catalog No. 50-753-4127 Supplier Julabo USA Inc Supplier No. 8981020
Bath Accessories, Julabo, M+R in-line Pt100 sensor with M16x1 male fittings, 1.5 m cable (measurement and control in external system). M16x1 male fittings with hose barb connections (barb connections are removable). For in-line Pt100 monitoring/contol from an external position in the flow circuit. For use with JULABO systems having external Pt100 capability.