Learn More
BioVendor, LLC Testosterone Mouse/Rat ELISA, 96 wells
SDP
Supplier:  BioVendor, LLC RTC001R
Encompass
Testosterone Mouse/Rat ELISA (96 wells). Immunoassay kit for quantitative measurement of testosterone in rat or mouse samples. Plasma, Serum: 10 uL/well. Calibration Range: 0.1–25 ng/mL. Limit of Detection: 0.066 ng/mL. Testosterone is the most important male sex hormone, it is responsible for genital development, beard growth, muscle development and general male characteristics. The measurement of serum or plasma levels is an index of leydig cell function and high or low values correlate well with hypo or hyper gonadism. In females small amounts of testosterone are produced by the adrenals and ovaries. High levels of testosterone in females indicates excessive androgen production and are found in progressive hirsutism and virilization, Cushing’s syndrome and a deficiency in one or more of the specific enzymes required for normal steroid biosynthesis.
Catalog No. 50-206-9287