Learn More
Lifeline Cell Technology M RENALEPI HU LIFELINE KT
SDP
Catalog No. 50311900
Encompass_Preferred
Includes RenaLife Basal Medium 485 mL, RenaLife LifeFactors Kit. Optimized for the culture of human renal epithelial cells. RenaLife supports the growth of these cells over a period of at least 15 population doublings.
Catalog No. 50-311-900 Supplier Lifeline Cell Technology Supplier No. LL0025
Includes RenaLife Basal Medium 485 mL, RenaLife LifeFactors Kit. Optimized for the culture of human renal epithelial cells. RenaLife supports the growth of these cells over a period of at least 15 population doublings.