Learn More
Bio IVT M RMONK CRY HEP 5 M CLLSV
Catalog No. 502310859
Encompass
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Rhesus monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial
Catalog No. 50-231-0859 Supplier Bio IVT Supplier No. M00315
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Rhesus monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial