Learn More
VEOLIA WTS M SERIES 12M CONSUMABLE KIT
Supplier:  VEOLIA WTS M9 MAINT
Encompass
includes: 4 persulfate reagents (150ml ea), 2 acid reagents, 2 uv lamps, 2 pump heads and 1 ion exchange resin bed.
Catalog No. NC2464455