Learn More
Clinical Diagnostic Solutions M-Series Cleaning Kit
Catalog No. 22999011
Change view
Click to view available options
For Use With (Equipment):
Medonic M Series
Catalog No. For Use With (Equipment)
22-999-011 Medonic M Series
1 options
Catalog No. 22-999-011 Supplier Clinical Diagnostic Solutions Supplier No. 501036
Add to Cart
Add to Cart
For use with Medonic M Series
Specifications
Description Clinical Diagnostic Solutions
For Use With (Equipment) Medonic M Series
Includes 450 mL Enzymatic Cleaner, 450 mL Hypochlorite Cleaner, 450 mL Detergent Cleaner
Intended Use Cleaning Kit for hematology systems