Learn More
Lifeline Cell Technology M SMC HU LIFELINE KT
SDP
Catalog No. 50311898
Encompass_Preferred
Includes VascuLife Basal Medium 475 mL, VascuLife SMC LifeFactors Kit. Optimized for the culture of human smooth muscle cells. Supports cell growth over a period of at least 15 population doublings. No antimicrobials and no phenol red.
Catalog No. 50-311-898 Supplier Lifeline Cell Technology Supplier No. LL0014
Includes VascuLife Basal Medium 475 mL, VascuLife SMC LifeFactors Kit. Optimized for the culture of human smooth muscle cells. Supports cell growth over a period of at least 15 population doublings. No antimicrobials and no phenol red.