Learn More
Zymo Research Corporation M-Solubilization Buffer (4.5 ml)
SDP
Catalog No. 502037119
Encompass_Preferred
The M-Solubilization Buffer is designed for use in the preparation of the CT Conversion Reagent in the EZ DNA Methylation-Direct Kit. The M-Solubilization Buffer facilitates the solubilization of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Direct Kits.
Catalog No. 50-203-7119 Supplier Zymo Research Corporation Supplier No. D50207
The M-Solubilization Buffer is designed for use in the preparation of the CT Conversion Reagent in the EZ DNA Methylation-Direct Kit. The M-Solubilization Buffer facilitates the solubilization of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Direct Kits.