Learn More
HARVEST RIGHT M SS FREEZE DRYER W/OIL FR PMP
Catalog No. NC3364743
Encompass_Preferred
  • Medium-sized freeze dryer for lab applications
  • Oil-free vacuum pump technology reduces maintenance
  • Full-color touchscreen display for easy monitoring
  • Adjustable freeze rates for precise control
  • 5 stainless steel trays for efficient drying
  • Holding capacity of 2 gallons of sublimated ice
  • Material capacity per batch of 2-4 gallons
  • Minimum collector temperature of -46 ºF
  • 115 VAC voltage for wall outlet compatibility
  • Manual or automated operation options
  • Recipe customization with 10 thermal treatments and 12 drying steps
  • Data logging for pressure and temperature throughout cycles
  • Ethernet connectivity for remote monitoring
  • Stainless steel construction for durability and cleanliness
  • 3-year limited warranty for peace of mind
Catalog No. NC3364743 Supplier HARVEST RIGHT Supplier No. HRFDMSC
Add to Cart
Add to Cart
Request a Quote
  • Medium-sized freeze dryer for lab applications
  • Oil-free vacuum pump technology reduces maintenance
  • Full-color touchscreen display for easy monitoring
  • Adjustable freeze rates for precise control
  • 5 stainless steel trays for efficient drying
  • Holding capacity of 2 gallons of sublimated ice
  • Material capacity per batch of 2-4 gallons
  • Minimum collector temperature of -46 ºF
  • 115 VAC voltage for wall outlet compatibility
  • Manual or automated operation options
  • Recipe customization with 10 thermal treatments and 12 drying steps
  • Data logging for pressure and temperature throughout cycles
  • Ethernet connectivity for remote monitoring
  • Stainless steel construction for durability and cleanliness
  • 3-year limited warranty for peace of mind