Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oak Ridge M-Style Sharps Container

Catalog No. 22730440
Change view
Click to view available options
Color:
Clear
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-730-440 Clear
22-730-441 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-730-440 Supplier Oak Ridge Supplier No. 0354110M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for in unsupervised patient areas

  • Provides safe disposal of syringes and needles
  • Transparent body allows for easy viewing
  • Expanded horizontal drop maximizes usable container volume for larger sharps

Specifications

Capacity (Metric) 4.7 L
Capacity (English) 5 qt.
Color Clear
For Use With (Application) Designed for use in unsupervised patient areas
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.