Learn More
Medchemexpress LLC m-Tolualdehyde | 620-23-5 | 99.6% | C8H8O | 5 G
SDP
Catalog No. 5000433310
Encompass_Preferred
m-Tolualdehyde is a natural product that can be isolated from *Ficus carica L.* and *Myosotis arvensis*. It possesses food protective effects and demonstrates acaricidal activities against *Tyrophagus putrescentiae*, with an LD50 of 1.97 μg/cm³.
  • It is a natural product.
  • It can be isolated from *Ficus carica L.* and *Myosotis arvensis*.
  • It has food protective effects.
  • It shows acaricidal activities for *Tyrophagus putrescentiae* with a LD50 of 1.97 μg/cm³.
Catalog No. 50-004-33310 Supplier Medchemexpress LLC Supplier No. HY780865G
m-Tolualdehyde is a natural product that can be isolated from *Ficus carica L.* and *Myosotis arvensis*. It possesses food protective effects and demonstrates acaricidal activities against *Tyrophagus putrescentiae*, with an LD50 of 1.97 μg/cm³.
  • It is a natural product.
  • It can be isolated from *Ficus carica L.* and *Myosotis arvensis*.
  • It has food protective effects.
  • It shows acaricidal activities for *Tyrophagus putrescentiae* with a LD50 of 1.97 μg/cm³.