Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Toluidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T030125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T030125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T030125G Supplier TCI America Supplier No. T030125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 638-03-9
Molecular Formula C7H10ClN
Molecular Weight (g/mol) 143.614
MDL Number MFCD00070607
InChI Key VDXZPXIEEVOGLV-UHFFFAOYSA-N
PubChem CID 522105
IUPAC Name 3-methylaniline;hydrochloride
SMILES CC1=CC(=CC=C1)N.Cl

Specifications

Melting Point 215°C
Color White
Quantity 25 g
Formula Weight 143.61
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material m-Toluidine Hydrochloride
EINECSNumber (3)-0186
RTECSNumber XU7175000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.