Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Toluoyl chloride, 99%

Catalog No. AC153415000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153415000 500 g Glass bottle
AC153410050 5 g Glass bottle
AC153411000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC153415000 Supplier Thermo Scientific Chemicals Supplier No. 153415000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1711-06-4 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-06-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000681
InChI Key YHOYYHYBFSYOSQ-UHFFFAOYSA-N
Synonym m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
PubChem CID 74375
IUPAC Name 3-methylbenzoyl chloride
SMILES CC1=CC=CC(=C1)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.1700g/mL
Boiling Point 86.0°C (5.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5475 to 1.5495
Linear Formula CH3C6H4COCl
Quantity 500 g
Beilstein 09, 477
Specific Gravity 1.17
Solubility Information Solubility in water: decomposes.
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Toluoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 216-976-8
RTECSNumber DM6644000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.