Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-tolylacetic acid (3-methylbenzeneacetic acid) | 621-36-3 | 99.9% | 150.18 g/mol | C9H10O2 | 10 G
SDP

Catalog No. 5000209999
Encompass_Preferred

m-tolylacetic acid (3-methylbenzeneacetic acid) is an organic chemical intermediate and analytical standard used in organic synthesis and research applications. It is supplied with high purity and characterized by its CAS number and molecular identifiers.

  • High purity (99.9%).
  • Molecular formula C9H10O2.
  • Molecular weight 150.18 g/mol.
  • CAS number 621-36-3.
  • Available in a 10 G package.
  • Intended for use as an organic synthesis intermediate and analytical standard.

Catalog No. 50-002-09999 Supplier Medchemexpress LLC Supplier No. HYW05350710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-tolylacetic acid (3-methylbenzeneacetic acid) is an organic chemical intermediate and analytical standard used in organic synthesis and research applications. It is supplied with high purity and characterized by its CAS number and molecular identifiers.

  • High purity (99.9%).
  • Molecular formula C9H10O2.
  • Molecular weight 150.18 g/mol.
  • CAS number 621-36-3.
  • Available in a 10 G package.
  • Intended for use as an organic synthesis intermediate and analytical standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.