Learn More
Medchemexpress LLC HY-W053507 500mg Medchemexpress, m-Tolylacetic acid CAS:621-36-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW053507500MG
Encompass_Preferred
Medchemexpress, HY-W053507 500mg m-Tolylacetic acid CAS:621-36-3 m-Tolylacetic acid (3-Methylbenzeneacetic acid) is a hydroaromatic dicarboxylic acids excreted in the urine as metabolite of tolueneacetic acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3490