Learn More
Bioss M. tuberculosis
SDP
Catalog No. 5000066735
Encompass_Preferred
M tuberculosis Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66735 Supplier Bioss Supplier No. BSM72295M
M tuberculosis Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide