Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-Tyramine hydrobromide | 38449-59-1 | 98.2% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY12897510MM/1ML

Encompass_Preferred

m-Tyramine hydrobromide is an endogenous trace amine neuromodulator that affects adrenergic and dopaminergic receptors.

  • Endogenous trace amine neuromodulator
  • Affects adrenergic and dopaminergic receptors
  • Appears as a solid, light yellow to yellow in color
  • Molecular weight: 218.09
  • Chemical formula: C8H12BrNO
  • Classified as alkaloids and phenols
  • Initial source is an endogenous metabolite

Catalog No. 50-003-39383


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.