Learn More
Medchemexpress LLC m-tyramine hydrobromide | 38449-59-1 | 98.5% | 218.09 g·mol-1 | C8H12BrNO | 5 MG
SDP
Catalog No. 5000264210
Encompass_Preferred
m-Tyramine hydrobromide is the hydrobromide salt of m-tyramine, an endogenous trace amine neuromodulator reported to interact with adrenergic and dopaminergic receptors. It is supplied for research and analytical applications, with reported analytical purity and supporting documentation for use as a reference standard.
  • Endogenous trace amine neuromodulator with adrenergic and dopaminergic activity.
  • Analytical grade with high purity suitable for research and standards.
  • Available in multiple small-scale pack sizes and as a solution in DMSO.
  • Supplied with certificate of analysis and RP-HPLC documentation.
  • Useful as a reference standard and biochemical research reagent.
Catalog No. 50-002-64210 Supplier Medchemexpress LLC Supplier No. HY1289755MG
m-Tyramine hydrobromide is the hydrobromide salt of m-tyramine, an endogenous trace amine neuromodulator reported to interact with adrenergic and dopaminergic receptors. It is supplied for research and analytical applications, with reported analytical purity and supporting documentation for use as a reference standard.
  • Endogenous trace amine neuromodulator with adrenergic and dopaminergic activity.
  • Analytical grade with high purity suitable for research and standards.
  • Available in multiple small-scale pack sizes and as a solution in DMSO.
  • Supplied with certificate of analysis and RP-HPLC documentation.
  • Useful as a reference standard and biochemical research reagent.