Learn More
Biotage Cap with Septum for Microwave Reaction Vials 100/Case
SDP
Catalog No. 502311209
Encompass
Biotage chemically resisitant caps and septa are the exact fit for Biotage microwave reaction vials. They are designed and tested to withstand pressures beyond 30 bar in a wide range of conditions. Manufactured from contaminant free microwave-safe materials. Reseal™ design means the septum reseals after it has been penetrated for repeated additions of reagents or in-situ sampling.This kit includes 100 each of aluminum caps and septa.
Catalog No. 50-231-1209 Supplier Biotage Supplier No. 352298
Biotage chemically resisitant caps and septa are the exact fit for Biotage microwave reaction vials. They are designed and tested to withstand pressures beyond 30 bar in a wide range of conditions. Manufactured from contaminant free microwave-safe materials. Reseal™ design means the septum reseals after it has been penetrated for repeated additions of reagents or in-situ sampling.This kit includes 100 each of aluminum caps and septa.