Learn More
Zymo Research Corporation M-Wash Buffer, 6 ml
SDP
Catalog No. 50444291
Encompass_Preferred
M-Wash Buffer (6 ml) For use with EZ-DNA Methylation™ bisulfite conversion kits. Sold individually or as part of a bisulfite conversion kit.
Catalog No. 50-444-291 Supplier Zymo Research Corporation Supplier No. D50014
M-Wash Buffer (6 ml) For use with EZ-DNA Methylation™ bisulfite conversion kits. Sold individually or as part of a bisulfite conversion kit.