Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Xylylenediamine

Catalog No. X1202100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X1202100G 100 g
X1202500G 500 g
X12025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. X1202100G Supplier Sigma Aldrich Supplier No. X1202100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • M-xylylenediamine can be used:
  • As a monomer for the synthesis of poly(vinylogous urethane) vitrimers
  • As a starting material for the synthesis of carbamate-protected multisubstituted bis-guanidine
  • For the synthesis of polyamides by catalytic dehydrogenation with diols

Specifications

CAS 1477-55-0
Density 1.032 g/mL (at 25°C (literature))
Boiling Point 265°C (745 mmHg)
Molecular Formula C8H12N2
Refractive Index n20/D 1.571 (literature)
Linear Formula C6H4(CH2NH2)2
MDL Number MFCD00008119
Quantity 100 g
Synonym 1,G-n3-G-nBenzenedimethanamine-m-XDA; 1,3-Bis(aminomethyl)benzene; alpha,alpha-Diamino-m-xylene
Molecular Weight (g/mol) 136.19
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.