Learn More
ATCC M059J; Malignant Glioblastoma; Human (Homo sapiens)
Catalog No. 502384750
Encompass
M059J is a glial cell isolated from the brain of a 33-year-old, male patient with malignant glioblastoma. These cell lines provide helpful model systems in which to study the role of DNA protein kinase in cellular and molecular processes involving DNA damage recognition and repair. It can be used in neuroscience research.
Catalog No. 50-238-4750 Supplier ATCC Supplier No. CRL2366
 Restricted Item
Add to Cart
Add to Cart
M059J is a glial cell isolated from the brain of a 33-year-old, male patient with malignant glioblastoma. These cell lines provide helpful model systems in which to study the role of DNA protein kinase in cellular and molecular processes involving DNA damage recognition and repair. It can be used in neuroscience research.