Learn More
ATCC M059K; Malignant Glioblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL2365
Encompass
M059K is a glial cell isolated from the brain of a 33-year-old, male patient with malignant glioblastoma. These cell lines provide helpful model systems in which to study the role of DNA protein kinase in cellular and molecular processes involving DNA damage recognition and repair. It can be used in neuroscience research.
Catalog No. 50-238-4153