Learn More
Teledyne ISCO, Inc M1 Class Single Piston Pump; Eccentric Drive; 0.0 - 40.0 Ml/Min. ; 500 Psi
Catalog No. 502119953 Shop All Teledyne ISCO Inc. Products
Encompass
M1 Class Single Piston Pump; Eccentric Drive; 0.0 - 40.0 mL/min. ; 500 psi; Stainless Steel Fluid Path; Automatic Piston Wash; without Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9953 Supplier Teledyne ISCO, Inc Supplier No. M1040SRX
M1 Class Single Piston Pump; Eccentric Drive; 0.0 - 40.0 mL/min. ; 500 psi; Stainless Steel Fluid Path; Automatic Piston Wash; without Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing