Learn More
Promocell GmbH PromoCell M1-Macrophage Generation Medium XF 250 ml, contains: C-28055B, C-39855, C-39894
Supplier:  Promocell GmbH C28055
Encompass
The PromoCell Macrophage Generation Media have been developed for the efficient generation of monocyte-derived macrophages (MDM) from freshly isolated peripheral blood monocytes or directly from PBMC as a starting material. The M1-Macrophage Generation Medium XF contains GM-CSF and allows for the generation of M1-polarized macrophages. PromoCells M1-Macrophage Generation Medium XF is a ready-to-use medium including cytokines for the directed differentiation of M1-like polarized MDM.The Macrophage Media XF are serum-free and xeno-free formulations for use with freshly isolated cells. Due to the utilization of exclusively synthetic, recombinant or plant-sourced materials, human serum albumin, purified from human plasma, is the only non-recombinant protein contained in this medium.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine macrophages.
Catalog No. NC0443997