Learn More
ATCC M1; Myeloid Leukemia; Mouse (Mus musculus)
Catalog No. 502384603
Encompass
M1 is a myeloblast cell isolated from a mouse with myeloid leukemia and can be used in immune system disorder research. The cell can be induced to become macrophage-like by a variety of stimulating agents including dexamethasone, LPS, and WEHI-3 conditioned medium.
Catalog No. 50-238-4603 Supplier ATCC Supplier No. TIB192
M1 is a myeloblast cell isolated from a mouse with myeloid leukemia and can be used in immune system disorder research. The cell can be induced to become macrophage-like by a variety of stimulating agents including dexamethasone, LPS, and WEHI-3 conditioned medium.