Description
- With calibration certificate
- Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
- Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
- Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Specifications
Specifications
| Product Type | Test Weight |
| Material | Stainless Steel |
| Case Material | Plastic |
| Weight (Metric) | 10 mg |
| Density | 7950 (± 140) kg/m3 |
| Shape | Sheet |
| Includes | With certificate |