Description
- Without calibration certificate
- For Standard Enviroments: Affordable quality weights for regular routine testing without the need for traceability
- Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Specifications
Specifications
| Product Type | Test Weight |
| Material | Stainless Steel |
| Case Material | Plastic |
| Weight (Metric) | 1 to 500 mg |
| Density | 7950 (± 140) kg/m3 |
| Includes | 12 Pieces; Without certificate |