Learn More
LifeSensors Tandem Ubiquitin Binding Entities (M1 TUBE) Magnetic 1mL
SDP
Catalog No. NC2285774
Encompass_Preferred
LifeSensors has developed TUBE (Tandem Ubiquitin Binding Entity) technology for the detection, characterization and isolation of polyubiquitylated proteins. These polyubiquitinated proteins can be detected in cells and tissue extracts. Our polyubiquitin capture reagents capitalize on the linkage of multiple ubiquitin interacting motifs (UIMs) to generate reagents with a high affinity for polyubiquitin. TUBEs have 100 to 1000-fold higher affinity for polyubiquitin chains compared to monomer ubiquitin binding domains (UBDs). TUBEs both stabilize and bind to ubiquitylated proteins, serving as an indispensable tool for ubiquitologists. Our M1 linear ubiquitin TUBE has high selectivity for M1 linear-linked polyubiquitin over the more common K48- and K63-linked polyubiquitin chains, making it a powerful tool for studying the biological consequences of this ubiquitin linkage type.
Catalog No. NC2285774 Supplier LifeSensors Supplier No. UM0406M1000
LifeSensors has developed TUBE (Tandem Ubiquitin Binding Entity) technology for the detection, characterization and isolation of polyubiquitylated proteins. These polyubiquitinated proteins can be detected in cells and tissue extracts. Our polyubiquitin capture reagents capitalize on the linkage of multiple ubiquitin interacting motifs (UIMs) to generate reagents with a high affinity for polyubiquitin. TUBEs have 100 to 1000-fold higher affinity for polyubiquitin chains compared to monomer ubiquitin binding domains (UBDs). TUBEs both stabilize and bind to ubiquitylated proteins, serving as an indispensable tool for ubiquitologists. Our M1 linear ubiquitin TUBE has high selectivity for M1 linear-linked polyubiquitin over the more common K48- and K63-linked polyubiquitin chains, making it a powerful tool for studying the biological consequences of this ubiquitin linkage type.