Learn More
Labtech Inc M10 Automated Concentrator. 10 positions supporting 50 & 200ml concentration cups simultaneously, 10 sensors for individual cup monitoring
Catalog No. 5000496182
Encompass_Preferred
M10 Automated Concentrator. 10 positions supporting 50 & 200ml concentration cups simultaneously, 10 sensors for individual cup monitoring
Catalog No. 50-004-96182 Supplier Labtech Inc Supplier No. MV10
M10 Automated Concentrator. 10 positions supporting 50 & 200ml concentration cups simultaneously, 10 sensors for individual cup monitoring