Learn More
Medchemexpress LLC HY-111547 5mg Medchemexpress, M1001 CAS:874590-32-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115475MG
Encompass_Preferred
Medchemexpress, HY-111547 5mg M1001 CAS:874590-32-6 M1001 is a weak agonist of HIF-2α, directly binds to the HIF-2α PAS-B domain, with a Kd of 667 nM. M1001 enhances the stabilities of HIF-2α-ARNT complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8913