Learn More
Chemglass Life Sciences M10 Adapter to 1/4" for Level Sensors
SDP
Supplier:  Chemglass Life Sciences CLS138092
Encompass
Used to install a 1/4" OD (6.35mm) level sensor into an M10 headplate port.Stainless steel components are manufactured from 316L with an electropolished surface finish of 20Ra or better.
Catalog No. 50-153-2116