Learn More
Maine Molecular Quality Controls Inc INTROL TB Panel M110
SDP
Supplier:  Maine Molecular Quality Controls Inc M110
Encompass
INTROL TB Panel M110 is intended for use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of M. tuberculosis (MTB) molecular assays. INTROL TB Panel M110 is formulated for use with in vitro assays that detect the presence of MTB and multi-drug resistant MTB (MDR-TB) mutations. INTROL TBWT-04, TBMDR1-04 and TBMDR2-04 of TB Panel M110 consist of non-infectious, synthetic MTB DNA encapsulated in chemically fixed and killed bacterial cells. The MTB DNA does not include the entire MTB genome but is comprised of the following MTB gene segments: IS6110, hsp65, 16S rRNA, 23S rRNA, inhA, katG, and rpoB. INTROL TB Panel M110 is comprised of four bottles, 3 mL each. Three bottles contain encapsulated MTB DNA, gene segments IS6110, hsp65, 16S rRNA, 23S rRNA, inhA, katG, and rpoB. The fourth bottle contains buffer and preservative only.
Catalog No. NC0950185