Learn More
Hach Company M12/M12 Ethernet Cable
Catalog No. 5000207054 Shop All Hach Company Products
Encompass
M12 connectors (2 pieces) for installation on standard Ethernet cable D-coded Cat 5 6-9 7 mm
Catalog No. 50-002-07054 Supplier Hach Company Supplier No. LXZ524.99.00010
M12 connectors (2 pieces) for installation on standard Ethernet cable D-coded Cat 5 6-9 7 mm