Learn More
Sino Biological Anti-M13 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11973MM05TP100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells. Then conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-7317