Learn More
TRIBIOSCIENCE INC M13 Phage/Bacteriophage qPCR Titer, 100 RXNs
SDP
Catalog No. NC3454593
Encompass_Preferred
Tribioscience’s M13 Phage qPCR FAST Titer Kit incorporates TribioScience FAST technology paradigm and Probe based qPCR technology for an accurate, one-step, and timesaving qPCR titration of M13. The probe is labeled with Fluorescence FAM. The entire procedure can be completed within 2-3 hours. With its time-saving 1-minute enzyme activation technology, specificity, and sensitivity, the performance of Tribioscience’s M13 Phase qPCR FAST Titer Kit is unmatched by similar kits currently on the market.
Catalog No. NC3454593 Supplier TRIBIOSCIENCE INC Supplier No. TBS4053
Tribioscience’s M13 Phage qPCR FAST Titer Kit incorporates TribioScience FAST technology paradigm and Probe based qPCR technology for an accurate, one-step, and timesaving qPCR titration of M13. The probe is labeled with Fluorescence FAM. The entire procedure can be completed within 2-3 hours. With its time-saving 1-minute enzyme activation technology, specificity, and sensitivity, the performance of Tribioscience’s M13 Phase qPCR FAST Titer Kit is unmatched by similar kits currently on the market.