Learn More
New England Biolabs, Inc. M13K07 Helper Phage – 1.8 ml
SDP
Catalog No. 50811731
Encompass
M13KO7 Helper Phage is used in conjunction with the phagemid of choice. In the presence of a phagemid bearing a wild-type M13 or f1 origin, single-stranded phagemid DNA is packaged preferentially and secreted into the culture medium. This allows easy production of single-stranded phagemid DNA.
Catalog No. 50-811-731 Supplier New England Biolabs, Inc. Supplier No. N0315S
M13KO7 Helper Phage is used in conjunction with the phagemid of choice. In the presence of a phagemid bearing a wild-type M13 or f1 origin, single-stranded phagemid DNA is packaged preferentially and secreted into the culture medium. This allows easy production of single-stranded phagemid DNA.