Learn More
BAYOU BIOLABS M13mp18 Single-Stranded Phage DNA, 500 μg, 1.00 μg/μL, Highly purified, Minimal contamination, 10 mM Tris-Cl, 1 mM EDTA, pH 8.0, Store at -20 °C

Supplier: BAYOU BIOLABS P107
This product is a highly purified M13mp18 single-stranded phage DNA that is essential for various molecular biology applications. It is engineered for efficient production of single-stranded DNA templates, making it suitable for dideoxy DNA sequencing, site-directed mutagenesis, and cloning. The product features a multiple cloning site within the lacZ' region, facilitating blue/white colony screening for recombinant selection, and is also suitable for advanced applications such as phage display and DNA origami.
- Type: Single-stranded phage DNA
- Purity: Highly purified, free of contaminants
- Quantity supplied: 500 μg
- Concentration: 1.00 μg/μL
- Storage temperature: -20 °C or below
- Quality control: Confirmed by electrophoresis
- Applications: DNA sequencing, cloning, site-directed mutagenesis
- Vector length: 7249 bp
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.