Learn More
New England Biolabs, Inc. M13mp18 RF I DNA – 10 µg
SDP
Catalog No. 50811788
Encompass
M13mp18 is the double-stranded, covalently closed, circular form of DNA derived from bacteriophage M13. This phage vector contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the -Galactosidase gene. When a fragment of DNA is inserted into one of these sites, the -Galactosidase gene is inactivated, providing selection for clones on the appropriate indicator plate.
Catalog No. 50-811-788 Supplier New England Biolabs, Inc. Supplier No. N4018S
M13mp18 is the double-stranded, covalently closed, circular form of DNA derived from bacteriophage M13. This phage vector contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the -Galactosidase gene. When a fragment of DNA is inserted into one of these sites, the -Galactosidase gene is inactivated, providing selection for clones on the appropriate indicator plate.