Learn More
Chemglass Life Sciences Chemglass M16 Blank Plug for Huber circulator fittings (CG-1969-M094)
SDP
Supplier:  Chemglass Life Sciences CG1969M094
Encompass
The Chemglass M16 Blank Plug (CG-1969-M094) is a stainless steel plug designed to seal unused M16-threaded ports on Huber circulator fittings and compatible fluid-transfer equipment. It provides a corrosion-resistant closure and is sold individually.
  • Made from stainless steel for corrosion resistance.
  • Thread size M16 for compatibility with M16 ports.
  • Used to blank or seal unused circulator fittings and ports.
  • Sold individually as a single unit.
  • No dimensional or pressure-rating data provided on the manufacturer page.
Catalog No. 50-273-3545