Learn More
Huber USA Adaptor
Catalog No. NC1124839
Encompass_Preferred
Adaptor M16x1 male thread to 1/2NPT female thread
Catalog No. NC1124839 Supplier Huber USA Supplier No. 6743
Adaptor M16x1 male thread to 1/2NPT female thread