Learn More
Chemglass Life Sciences Chemglass adapter, M16 x 1 male to 1/4 in female NPT - stainless steel (CG-1969-M204)
SDP
Catalog No. 502733543
Encompass
This stainless steel adapter converts an M16 x 1 male circulator port to a 1/4 in female NPT connection, providing a secure transition between metric Huber circulator fittings and NPT-threaded tubing or components.
  • Made from stainless steel.
  • M16 x 1 male to 1/4 in female NPT connection.
  • Compatible with Huber circulator fittings.
  • Sold individually.
Catalog No. 50-273-3543 Supplier Chemglass Life Sciences Supplier No. CG1969M204
This stainless steel adapter converts an M16 x 1 male circulator port to a 1/4 in female NPT connection, providing a secure transition between metric Huber circulator fittings and NPT-threaded tubing or components.
  • Made from stainless steel.
  • M16 x 1 male to 1/4 in female NPT connection.
  • Compatible with Huber circulator fittings.
  • Sold individually.