Learn More
Chemglass Life Sciences Adapter, M16 x 1 Male to 1/4 in Male NPT, Stainless Steel (for Huber circulator fittings)
SDP
Catalog No. 502733544
Encompass
Chemglass stainless steel adapter that converts an M16 x 1 male thread to a 1/4 in male NPT connection for laboratory circulator and fluid transfer applications. It is commonly used with circulator fittings to connect equipment and tubing.
  • Converts M16 x 1 male thread to 1/4 in male NPT connection.
  • Made from stainless steel for corrosion resistance and durability.
  • Compatible with laboratory circulator fittings and fluid transfer systems.
  • Compact form factor for easy installation in tight plumbing setups.
  • Sold individually for replacement or spare parts needs.
Catalog No. 50-273-3544 Supplier Chemglass Life Sciences Supplier No. CG1969M104
Chemglass stainless steel adapter that converts an M16 x 1 male thread to a 1/4 in male NPT connection for laboratory circulator and fluid transfer applications. It is commonly used with circulator fittings to connect equipment and tubing.
  • Converts M16 x 1 male thread to 1/4 in male NPT connection.
  • Made from stainless steel for corrosion resistance and durability.
  • Compatible with laboratory circulator fittings and fluid transfer systems.
  • Compact form factor for easy installation in tight plumbing setups.
  • Sold individually for replacement or spare parts needs.