Learn More
Chemglass Life Sciences Blind Stoppers for Bioreactors M18
SDP
Catalog No. NC1779817
Encompass
Stoppers can be used to close off any of the unused openings in the head plate. The M18, M27, G3/4 and M30 are threaded into the ports from the outside. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.
This item is not returnable. View return policy
Catalog No. NC1779817 Supplier Chemglass Life Sciences Supplier No. CLS138025
Stoppers can be used to close off any of the unused openings in the head plate. The M18, M27, G3/4 and M30 are threaded into the ports from the outside. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.